missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, Reagent Grade, ≥99%, Solstice

Product Code. 15687790 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
4 x 2.5 L
6 x 1 L
Packaging:
Poly Bottle
Unit Size:
1 L
2.5 L
5 L
500mL
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15687790 2.5 L Poly Bottle 2.5 L
15627800 500 mL Poly Bottle 500mL
15677790 1 L Poly Bottle 1 L
15637800 5 L Poly Bottle 5 L
15647800 6 x 1 L Poly Bottle Pack of 6
15617800 4 x 2.5 L Poly Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15687790 Supplier Fluka Supplier No. D1585502.5L
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 0.944 g/mL(lit.)
Quantity 2.5 L
Explosion Limit 0.152
Flash Point 57.5°C
Ignition Point 833°F
Packaging Poly Bottle
Linear Formula HCON(CH3)2
Vapor Pressure 2.7 mmHg (20°C)
UN Number UN2265
Water max. 0.1%
Beilstein 605365
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
Boiling Point 152°C to 153°C
Formula Weight 73.10
Percent Purity >99%
Grade Reagent
Chemical Name or Material N,N-Dimethylformamide, Reagent Grade, ≥99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312+H332-Harmful in contact with skin or if inhaled
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.