missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylglycine Hydrochloride, TRC

Product Code. 30370114
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370114 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370114 Supplier Toronto Research Chemicals Supplier No. TRCD47806010G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethylglycine Hydrochloride
CAS 2491-06-7
Molecular Formula C4 H9 N O2 . HCl
Quantity 10 g
Synonym Glycine, N,N-dimethyl-, hydrochloride (1:1), Glycine, N,N-dimethyl-, hydrochloride (6CI,8CI,9CI), (Carboxymethyl)dimethylazanium chloride, (Dimethylamino)acetic acid hydrochloride, 2-(Dimethylamino)acetic acid hydrochloride, Dimethylglycine hydrochloride, N,N-Dimethylaminoacetic acid hydrochloric acid, N,N-Dimethylglycine hydrochloride, N,N-Dimethylglycine monohydrochloride
InChI Formula InChI=1S/C4H9NO2.ClH/c1-5(2)3-4(6)7;/h3H2,1-2H3,(H,6,7);1H
SMILES Cl.CN(C)CC(=O)O
Molecular Weight (g/mol) 139.58
Formula Weight 139.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.