missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylglycine, TRC

Product Code. 30370395
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30370395 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30370395

Brand: Toronto Research Chemicals TRCD47308025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethylglycine
CAS 1118-68-9
Molecular Formula C4H9NO2
Quantity 25 g
Synonym (Dimethylamino)acetic Acid, 2-(Dimethylamino)acetic Acid, 2-(N,N-Dimethylamino)acetic Acid, 46: PN: WO2008005407 PAGE: 5 claimed protein, DMG, N,N-Dimethylaminoacetic Acid, N-Methylsarcosine, Dimethylglycine
InChI Formula InChI=1S/C4H9NO2/c1-5(2)3-4(6)7/h3H2,1-2H3,(H,6,7)
SMILES CN(C)CC(=O)O
Molecular Weight (g/mol) 103.12
Formula Weight 103.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.