missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylhexadecylamine-N-Oxide, TRC

Product Code. 30359122
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359122 1 g 1g
30352242 100 mg 100mg
30352773 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359122 Supplier Toronto Research Chemicals Supplier No. TRCD4059201G
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7128-91-8
Molecular Formula C18H39NO
Molecular Weight (g/mol) 285.52
Synonym Admox 16, Ammonyx CO, Aromox DM 16, Aromox DM 16D, Aromox DM 16D-M, Barlox 16, Barlox 16S, Cetamine Oxide, Cetyldimethylamine oxide, Conco XAC, Dimethylcetylamine oxide, Dimethylhexadecylamine oxide, Dimethylpalmitylamine oxide, Hexadecyldimethylamine N-oxide, Hexadecyldimethylamine oxide, N,N-Dimethyl-1-hexadecanamine oxide, N,N-Dimethyl-N-hexadecylamine N-oxide, N,N-Dimethyl-n-hexadecylamine oxide, N,N-Dimethylhexadecylamine oxide, N-Cetyldimethylamine Oxide, N-Hexadecyl-N,N-dimethylamine oxide, N-Palmityl-N,N-dimethylamine oxide, Palmityldimethylamine oxide
IUPAC Name N,N-dimethylhexadecan-1-amine oxide
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H39NO/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(2,3)20/h4-18H2,1-3H3
Formula Weight 285.3032
Chemical Name or Material N,N-Dimethylhexadecylamine-N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.