missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylhexanamide, 98%

Product Code. 11490398
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11490398

Brand: Thermo Scientific Alfa Aesar B24562.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5830-30-8 | C8H17NO | 143.23 g/mol

N,N-Dimethylhexanamide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylhexanamide is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5830-30-8
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD00043671
InChI Key OAERLTPBKQBWHJ-UHFFFAOYSA-N
Synonym hexanamide, n,n-dimethyl, n,n-dimethylcaproamide, n,n-dimethylhexanoamide, nn-dimethylhexanamide, nn-dimethylcaproamide, n,n-dirnethylhexanamide, n,n-dimethyl-hexanamide, hexanamide,n,n-dimethyl, acmc-1atq3, n,n-dimethylhexanamide, tech
PubChem CID 22084
IUPAC Name N,N-dimethylhexanamide
SMILES CCCCCC(=O)N(C)C

Specifications

Density 0.89
Boiling Point 80°C (1 mmHg)
Refractive Index 1.449
Quantity 5 g
Beilstein 1747074
Solubility Information Not miscible or difficult to mix.
Formula Weight 143.23
Percent Purity 98%
Chemical Name or Material N,N-Dimethylhexanamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.