missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylpropionamide, 98+%

Product Code. 15482239
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482239 25 g 25g
15492239 100 g 100g
15402249 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15482239 Supplier Thermo Scientific Alfa Aesar Supplier No. H64850.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 758-96-3 | C5H11NO | 101.149 g/mol

N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.

Solubility
Soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 758-96-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00009301
InChI Key MBHINSULENHCMF-UHFFFAOYSA-N
Synonym n,n-dimethylpropionamide, propanamide, n,n-dimethyl, dimethylamide of propionic acid, propionamide, n,n-dimethyl, n,n-dimethyl propionamide, acmc-209p0o, propionic acid dimethylamide, 4-04-00-00184 beilstein handbook reference, ksc494e1j
PubChem CID 12965
IUPAC Name N,N-dimethylpropanamide
SMILES CCC(=O)N(C)C

Specifications

Melting Point -45°C (literature)
Color Colorless
Density 0.920 g/mL
Boiling Point 174°C to 175°C (literature)
Flash Point 62°C (144°F)
Linear Formula CH3CH2CON(CH3)2
Refractive Index 1.44
Quantity 25 g
Beilstein 1740719
Solubility Information Soluble in water.
Formula Weight 101.15
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material N,N-Dimethylpropionamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.