missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenylbenzidine, 97%

Product Code. 10763944 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
This item is not returnable. View return policy

Product Code. 10763944

Brand: Thermo Scientific 117210500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 531-91-9 | C24H20N2 | 336.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-91-9
Molecular Formula C24H20N2
Molecular Weight (g/mol) 336.44
MDL Number MFCD00003016
InChI Key FDRNXKXKFNHNCA-UHFFFAOYSA-N
Synonym n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine
PubChem CID 68280
IUPAC Name 4-(4-anilinophenyl)-N-phenylaniline
SMILES N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 246°C to 250°C
Color Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Beilstein 13, 223
Merck Index 15, 3349
Formula Weight 336.42
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material N, N'-Diphenylbenzidine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.