missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenylformamidine, TRC

Product Code. 30382840
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382840 25 g
30382261 50 g
30385373 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382840 Supplier Toronto Research Chemicals Supplier No. TRCD49100025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-15-1
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.25
Synonym N,N'-Diphenylmethanimidamide, 1,3-Diphenyl-1,3-diaza-1-propene, Diphenylformamidine
SMILES N(\C=N\c1ccccc1)c2ccccc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C13H12N2/c1-3-7-12(8-4-1)14-11-15-13-9-5-2-6-10-13/h1-11H,(H,14,15)
Chemical Name or Material N,N'-Diphenylformamidine
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.