missing translation for 'onlineSavingsMsg'
Learn More

N,N′-Disuccinimidyl Carbonate, TRC

Product Code. 30382391
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382391 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382391 Supplier Toronto Research Chemicals Supplier No. TRCD4935405G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Disuccinimidyl Carbonate
CAS 74124-79-1
Molecular Formula C9 H8 N2 O7
Quantity 5 g
Synonym Carbonic acid, bis(2,5-dioxo-1-pyrrolidinyl) ester (ACI), 2,5-Pyrrolidinedione, 1,1'-[carbonylbis(oxy)]bis- (9CI), 1-[[[(2,5-Dioxo-1-pyrrolidinyl)oxy]carbonyl]oxy]-2,5-pyrrolidinedione, Bis(2,5-dioxopyrrolidin-1-yl) carbonate, Bis(N-succinimidyl) carbonate, Bis(succinimidyl) carbonate, Carbonic acid bis(2,5-dioxopyrrolidin-1-yl) ester, Carbonic acid disuccinimidyl ester, Di(2,5-dioxopyrrolidin-1-yl) carbonate, Di(N-succinimidyl) carbonate, Disuccimidyl carbonate, Disuccinimide carbonate, Disuccinimidyl carbonate, DSC, N,N-Disuccinimidyl carbonate, N,N'-Disuccinimido carbonate, N,N'-Disuccinimidyl carbonate
InChI Formula InChI=1S/C9H8N2O7/c12-5-1-2-6(13)10(5)17-9(16)18-11-7(14)3-4-8(11)15/h1-4H2
SMILES O=C(ON1C(=O)CCC1=O)ON2C(=O)CCC2=O
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) carbonate
Molecular Weight (g/mol) 256.17
Formula Weight 256.0332
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.