missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Methylene-Bis-Acrylamide, Ultra Pure, 50g, MP Biomedicals

Product Code. 11401542 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401542 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11401542 Supplier MP Biomedicals Supplier No. 04800173

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

110-26-9 Purity: 99.9% (290nm): <<0.2 Acrylic acid content: <<0.02% High purity cross-linker used for precise, critical PAGE gels. May be used in UV scanning gels

Specifications

CAS 110-26-9
Quantity 50 g
Molecular Formula C7H10N2O2
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
SMILES C=CC(=O)NCNC(=O)C=C
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
Molecular Weight (g/mol) 154.169
PubChem CID 8041
Chemical Name or Material N, N«-Methylene-Bis-Acrylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.