missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N'-Methylenebisacrylamide, 2% soln.

Product Code. 15415539
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415539 500 mL 500mL
15425539 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415539 Supplier Thermo Scientific Chemicals Supplier No. J63265.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol

Bis-acrylamide cross-links with acrylamide to form polyacrylamide gels for electrophoresis. Changing the ratio of bis-acrylamide to acrylamide controls the pore size of polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Bis-acrylamide cross-links with acrylamide to form polyacrylamide gels for electrophoresis
  • Changing the ratio of bis-acrylamide to acrylamide controls the pore size of polyacrylamide gels
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00008625
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Color Colorless
Quantity 500 mL
Formula Weight 154.17
Concentration or Composition (by Analyte or Components) 2% soln.
Physical Form Liquid
Chemical Name or Material N,N'-Methylenebisacrylamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.