missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Methylenebisacrylamide, 97%

Product Code. 11351677
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11351677 500 g 500g
11341677 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351677 Supplier Thermo Scientific Alfa Aesar Supplier No. L04694.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol

N,N'-Methylenebisacrylamide is used as an intermediate and cross-linking agent utilized in the preparation of polymers like polyacrylamide. It is also used in biochemistry for the preparation of polyacrylamide gels, which is useful for the electrophoretic separation of nucleic acids and proteins. Further, it is employed in the synthesis of acrylic resin and adhesives. In addition to this, it is used as a plugging agent in oilfield drilling and construction grouting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Methylenebisacrylamide is used as an intermediate and cross-linking agent utilized in the preparation of polymers like polyacrylamide. It is also used in biochemistry for the preparation of polyacrylamide gels, which is useful for the electrophoretic separation of nucleic acids and proteins. Further, it is employed in the synthesis of acrylic resin and adhesives. In addition to this, it is used as a plugging agent in oilfield drilling and construction grouting.

Solubility
Soluble in water, ethanol and methanol.

Notes
Air and light sensitive. Store in a cool place. Incompatible with acids, bases, oxidizing agents, reducing agents, iron, iron salts, copper, aluminum, brass and free radical initiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00008625
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Melting Point >300°C
Density 1.235
Quantity 500 g
Beilstein 1706297
Solubility Information Soluble in water,ethanol and methanol.
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material N,N'-Methylenebisacrylamide
compliance-icons compliance-icons
Product Identifier
  • N,N'-Methylenebisacrylamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Methylene diacrylamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.