missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetraacetylethylenediamine, 99%, Thermo Scientific™

Product Code. 15212530
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15212530 50 g 50g
11306906 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15212530 Supplier Thermo Scientific Alfa Aesar Supplier No. L04353.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N,N,N′,N′-Tetraacetylethylenediamine is used as a peroxide bleach activator in household detergents and for paper pulp. It also serves as an important component of laundry detergents and bleaches, where it is used as an activator for active oxygen bleaching agents such as sodium perborate, sodium percarbonate, sodium perphosphate and sodium persulfate. It reacts with the perhydroxyl anion HO2- in the presence of aqueous alkaline solution to prepare triacetylethylenediamine and diacetylethylenediamine with the release of peracetic acid, which is a fast acting bleaching agent.

Solubility
Soluble in chloroform and methylenechloride. Slightly soluble in water and acetone.

Notes
Incompatible with bases and metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10543-57-4
Molecular Formula C10H16N2O4
Molecular Weight (g/mol) 228.248
MDL Number MFCD00014967
InChI Key BGRWYDHXPHLNKA-UHFFFAOYSA-N
Synonym tetraacetylethylenediamine, n,n,n',n'-tetraacetylethylenediamine, taed, tetracetylethylenediamine, acetamide, n,n'-1,2-ethanediylbis n-acetyl, n,n'-ethylenebis n-acetylacetamide, unii-p411ed0n2b, n,n'-ethane-1,2-diyl bis n-acetylacetamide, n-acetyl-n-2-diacetylamino ethyl acetamide
PubChem CID 66347
IUPAC Name N-acetyl-N-[2-(diacetylamino)ethyl]acetamide
SMILES CC(=O)N(CCN(C(=O)C)C(=O)C)C(=O)C

Specifications

Melting Point 149°C to 153°C
Quantity 50 g
Beilstein 1795711
Merck Index 14,9028
Solubility Information Soluble in chloroform and methylenechloride. Slightly soluble in water and acetone.
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material N,N,N',N'-Tetraacetylethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.