missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Product Code. 11995261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11995261 100 g Glass Bottle 100g
10327640 5 g Glass Bottle 5g
10215990 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11995261 Supplier Thermo Scientific Acros Supplier No. 170791000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-22-1 | C10H16N2 | 164.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-22-1
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.25
InChI Key CJAOGUFAAWZWNI-UHFFFAOYSA-N
Synonym wurster's blue, wurster's reagent, n,n,n',n'-tetramethyl-p-phenylenediamine, n,n,n',n'-tetramethyl-1,4-phenylenediamine, tetramethyl-p-phenylenediamine, 1,4-bis dimethylamino benzene, p-bis dimethylamino benzene, 1,4-benzenediamine, n,n,n',n'-tetramethyl, benzene, 1,4-bis dimethylamino, unii-p4p3ac32zb
PubChem CID 7490
IUPAC Name 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine
SMILES CN(C)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 48.0°C to 52.0°C
Color Beige-Brown to Brown
Boiling Point 260.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula C6H4[N(CH3)2]2
Quantity 100 g
Beilstein 13,74
Merck Index 15,9376
Solubility Information Solubility in water: slightly in cold, more in hot water. Other solubilities: soluble in alcohols, chloroform, ether
Formula Weight 164.25
Percent Purity 98%
Physical Form Crystalline Flakes
Chemical Name or Material N,N,N',N'-Tetramethyl-p-phenylenediamine
Show More Show Less
compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethyl-p-phenylenediamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.