missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyldiaminomethane, 99%

Product Code. 10456770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10456770 100 mL Glass bottle 100mL
10133620 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10456770 Supplier Thermo Scientific Acros Supplier No. 138401000
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51-80-9 | C5H14N2 | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-80-9
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008328
InChI Key VGIVLIHKENZQHQ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyldiaminomethane, bis dimethylamino methane, methanediamine, n,n,n',n'-tetramethyl, tetramethylmethylenediamine, methylenebis dimethylamine, n,n,n',n'-tetramethylmethylenediamine, unii-z870i525ks, methylenediamine, n,n,n',n'-tetramethyl, dimethylamino methyl dimethylamine, dimethyl dimethylamino methyl amine
PubChem CID 5829
IUPAC Name N,N,N',N'-tetramethylmethanediamine
SMILES CN(C)CN(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.7400g/mL
Boiling Point 85.0°C
Flash Point −12°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCH2N(CH3)2
Refractive Index 1.3995 to 1.4015
Quantity 100 mL
Beilstein 04, 54
Fieser 03,21; 07,359
Specific Gravity 0.74
Formula Weight 102.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethyldiaminomethane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethyldiaminomethane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.