missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade, MP Biomedicals

Product Code. 11471612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471612 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11471612 Supplier MP Biomedicals Supplier No. 04805615
  • ONLINE EXCLUSIVE PRICE
    128.00 €
    120.75 € / 50mL
    Save 7.25 €
    5% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

110-18-9 (TEMED) Purity: >99% Polymerization accelerator for polyacrylamide gel formation

Specifications

CAS 110-18-9
Molecular Formula C6H16N2
Quantity 50 mL
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILES CN(C)CCN(C)C
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
Molecular Weight (g/mol) 116.208
PubChem CID 8037
ChEBI CHEBI:32850
Percent Purity ≥99%
Grade Electrophoresis
Chemical Name or Material N,N,N«,N«-Tetramethyl-Ethylenediamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • TEMED, Electrophoresis Grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: N,N,N',N'-tetramethylethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.