missing translation for 'onlineSavingsMsg'
Learn More

N,N,Nft.,Nft.-Tetramethylethylenediamine, Molecular biology reagent grade, MP Biomedicals™

Product Code. 11496491 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496491 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496491 Supplier MP Biomedicals Supplier No. 0219401950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N,N,N',N'-Tetramethylethylenediamin is a tertiary amine base

TRUSTED_SUSTAINABILITY

Specifications

CAS 110-18-9
Density 0.775g/mL at 20°C
Assay Percent Range Approx. 99%
Molecular Formula C6H16N2
MDL Number MFCD00008335
Quantity 50 mL
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILES CN(C)CCN(C)C
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
Molecular Weight (g/mol) 116.208
PubChem CID 8037
ChEBI CHEBI:32850
Formula Weight 116.2
Grade Molecular Biology Reagent
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.