missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Thiodiphthalimide (>90%), TRC

Product Code. 30474790
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474790 1 g -
30473412 500 mg 500mg
30482049 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474790 Supplier Toronto Research Chemicals Supplier No. TRCN5974001G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7764-29-6
Molecular Formula C16 H8 N2 O4 S
Molecular Weight (g/mol) 324.31
Synonym 2,2'-thiobis(isoindoline-1,3-dione), N,N'-thiodi Phthalimide, 2,2'-Thiobis[1H-isoindole-1,3(2H)-dione], N,N'-Thiobisphthalimide
IUPAC Name 2-(1,3-dioxoisoindol-2-yl)sulfanylisoindole-1,3-dione
SMILES O=C1N(SN2C(=O)c3ccccc3C2=O)C(=O)c4ccccc14

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H8N2O4S/c19-13-9-5-1-2-6-10(9)14(20)17(13)23-18-15(21)11-7-3-4-8-12(11)16(18)22/h1-8H
Formula Weight 324.0205
Chemical Name or Material N,N'-Thiodiphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.