missing translation for 'onlineSavingsMsg'
Learn More

N-Nervonoyl-D-sphingosine, TRC

Product Code. 30179539
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179539 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30179539 Supplier Toronto Research Chemicals Supplier No. TRCN39010010MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 54164-50-0
Molecular Formula C42 H81 N O3
Quantity 10 mg
Synonym (15Z)-N-[(1S,2R,3E)-2-Hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]-15-tetracosenamide, 15-Tetracosenamide, N-[(1S,2R,3E)-2-hydroxy-1-(hydroxymethyl)-3-heptadecenyl]-, (15Z)- (9CI), 15-Tetracosenamide, N-[2-hydroxy-1-(hydroxymethyl)-3-heptadecenyl]-, [R-[R*,S*-(E,Z)]]-, (15Z)-N-[(1S,2R,3E)-2-Hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]-15-tetracosenamide, C24:1-Ceramide, D-erythro-1,3-Dihydroxy-2-(cis-15-tetracosenoylamido)-trans-4-octadecene, N-Nervonoyl-D-sphingosine, N-cis-15-Tetracosenoyl-C18-sphingosine
InChI Formula InChI=1S/C42H81NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-38-42(46)43-40(39-44)41(45)37-35-33-31-29-27-25-16-14-12-10-8-6-4-2/h17-18,35,37,40-41,44-45H,3-16,19-34,36,38-39H2,1-2H3,(H,43,46)/b18-17-,37-35+/t40-,41+/m0/s1
SMILES CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(=O)CCCCCCCCCCCCC\C=C/CCCCCCCC
IUPAC Name (Z)-N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]tetracos-15-enamide
Molecular Weight (g/mol) 648.1
Formula Weight 647.6216
Chemical Name or Material Nervonyl-D-sphingosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.