missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Desloratadine-d4, TRC

Product Code. 30477716
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477716 25 mg -
30469731 5 mg -
30465552 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30477716 Supplier Toronto Research Chemicals Supplier No. TRCN52537225MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2733579-31-0
Molecular Formula C19H14D4ClN3O
Molecular Weight (g/mol) 343.85
Synonym 8-Chloro-6,11-dihydro-11-(1-nitroso-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine-d4
IUPAC Name 8-chloro-11-(1-nitrosopiperidin-4-ylidene-2,2,6,6-d4)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
SMILES O=NN(C([2H])([2H])C/1)C([2H])([2H])CC1=C2C3=C(C=CC=N3)CCC4=C/2C=CC(Cl)=C4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H18ClN3O/c20-16-5-6-17-15(12-16)4-3-14-2-1-9-21-19(14)18(17)13-7-10-23(22-24)11-8-13/h1-2,5-6,9,12H,3-4,7-8,10-11H2/i10D2,11D2
Formula Weight 343.14
Percent Purity >95%
Chemical Name or Material N-Nitroso Desloratadine-d4
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.