missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Desmethyl-spinosyn Factor A, TRC

Product Code. 30427332
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427332 25 mg 25mg
30426176 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30427332 Supplier Toronto Research Chemicals Supplier No. TRCN70359525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso Desmethyl-spinosyn Factor A
CAS 252575-85-2
Molecular Formula C40H62N2O11
Quantity 25 mg
Synonym 1H-as-Indaceno[3,2-d]oxacyclododecin-7,15-dione','2-[(6-deoxy-2,3,4-tri-O-methyl-α-L-mannopyranosyl)oxy]-9-ethyl-2,3,3a,5a,5b,6,9,10,11,12,13,14,16a,16b-tetradecahydro-14-methyl-13-[[(2R,5S,6R)-tetrahydro-6-methyl-5-(methylnitrosoamino)-2H-pyran-2-yl]oxy]-','(2R,3aS,5aR,5bS,9S,13S,14R,16aS,16bR)- (9CI','ACI), (2R,3aS,5aR,5bS,9S,13S,14R,16aS,16bR)-2-[(6-Deoxy-2,3,4-tri-O-methyl-α-L-mannopyranosyl)oxy]-9-ethyl-2,3,3a,5a,5b,6,9,10,11,12,13,14,16a,16b-tetradecahydro-14-methyl-13-[[(2R,5S,6R)-tetrahydro-6-methyl-5-(methylnitrosoamino)-2H-pyran-2-yl]oxy]-1H-as-indaceno[3,2-d]oxacyclododecin-7,15-dione (ACI)
InChI Formula InChI=1S/C40H62N2O11/c1-9-25-11-10-12-33(53-35-16-15-32(22(3)49-35)42(5)41-45)21(2)36(44)31-19-29-27(30(31)20-34(43)51-25)14-13-24-17-26(18-28(24)29)52-40-39(48-8)38(47-7)37(46-6)23(4)50-40/h13-14,19,21-30,32-33,35,37-40H,9-12,15-18,20H2,1-8H3/t21-,22-,23-,24?,25+,26-,27?,28?,29?,30?,32+,33+,35+,37-,38+,39+,40-/m1/s1
SMILES O=C([C@@H]([C@H](CCC[C@@H](O1)CC)O[C@@H]2O[C@@H]([C@H](CC2)N(C)N=O)C)C)C3=CC(C(C=CC4C5)C3CC1=O)C4C[C@@H]5O[C@@H]6[C@@H](OC)[C@@H](OC)[C@H](OC)[C@@H](C)O6
IUPAC Name N-((2R,3S,6R)-6-(((2R,9S,13S,14R)-9-ethyl-14-methyl-7,15-dioxo-2-(((2S,3S,4S,5R,6R)-3,4,5-trimethoxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2,3,3a,5a,5b,6,7,9,10,11,12,13,14,15,16a,16b-hexadecahydro-1H-as-indaceno[3,2-d][1]oxacyclododecin-13-yl)oxy)-2-methyltetrahydro-2H-pyran-3-yl)-N-methylnitrous amide
Molecular Weight (g/mol) 746.94
Formula Weight 746.44
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.