missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Hydrochlorothiazide, TRC

Product Code. 30003740
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003740 250 mg 250mg
17953195 25 mg 25mg
17910915 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003740 Supplier Toronto Research Chemicals Supplier No. TRCN542055250MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63779-86-2
Molecular Formula C7 H7 Cl N4 O5 S2
Molecular Weight (g/mol) 326.737
Synonym 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 6-chloro-3,4-dihydro-4-nitroso-, 1,1-dioxide, 4-Nitrosohydrochlorothiazide
IUPAC Name 6-chloro-4-nitroso-1,1-dioxo-2,3-dihydro-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)c1cc2c(cc1Cl)N(CNS2(=O)=O)N=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H7ClN4O5S2/c8-4-1-5-7(2-6(4)18(9,14)15)19(16,17)10-3-12(5)11-13/h1-2,10H,3H2,(H2,9,14,15)
Formula Weight 325.955
Percent Purity >95%
Chemical Name or Material 4-Nitrosohydrochlorothiazide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.