missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N-methyl-N-dodecylamine, TRC

Product Code. 30041167
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041167 10 mg 10mg
30040776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30041167 Supplier Toronto Research Chemicals Supplier No. TRCN52993510MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55090-44-3
Molecular Formula C13H28N2O
Molecular Weight (g/mol) 228.37
Synonym 1-Dodecanamine, N-methyl-N-nitroso- (9CI, ACI), N-Methyl-N-nitroso-1-dodecanamine (ACI), MDNA, Methyldodecylnitrosamine, N-Methyl-N-dodecylnitrosamine, N-Nitroso-N-methyl-N-dodecylamine, N-Nitrosomethyldodecylamine, Nitrosomethyldodecylamine, N-Nitroso-N-methyldodecylamine
IUPAC Name N-dodecyl-N-methylnitrous amide
SMILES CCCCCCCCCCCCN(C)N=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H28N2O/c1-3-4-5-6-7-8-9-10-11-12-13-15(2)14-16/h3-13H2,1-2H3
Formula Weight 228.2202
Chemical Name or Material N-Nitroso-N-methyl-N-dodecylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.