missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N,N-di-(7-methyloctyl)amine, TRC

Product Code. 30009191
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009191 2.5 mg 2.5mg
30004097 1 mg 1mg
17950685 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009191 Supplier Toronto Research Chemicals Supplier No. TRCN5257002.5MG
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 643014-99-7
Molecular Formula C18 H38 N2 O
Molecular Weight (g/mol) 298.51
IUPAC Name N,N-bis(7-methyloctyl)nitrous amide
SMILES CC(C)CCCCCCN(CCCCCCC(C)C)N=O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C18H38N2O/c1-17(2)13-9-5-7-11-15-20(19-21)16-12-8-6-10-14-18(3)4/h17-18H,5-16H2,1-4H3
Formula Weight 298.2984
Percent Purity >95%
Chemical Name or Material N-Nitroso-N,N-di-(7-methyloctyl)amine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.