missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosodiethyl-d10-amine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-Nitroso-diethylamine-d10 |
| CAS | 1219794-54-3 |
| Molecular Formula | C4 2H10 N2 O |
| Quantity | 0.05 g |
| Synonym | N-Nitroso-diethylamine D10, N-(Ethyl-d5)-N-nitrosoethan-d5-amine, N-Nitroso-bis(pentadeuterioethyl)amine |
| InChI Formula | InChI=1 S/C4H10N2O/c1-3-6(4-2)5-7/h3-4H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])N(N=O)C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N-bis(1,1,2,2,2-pentadeuterioethyl)nitrous amide |
| Molecular Weight (g/mol) | 112.2 |
| Formula Weight | 112.1421 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction