missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodiethylamine (1mg/mL in Methanol), TRC

Product Code. 30009907
Change view
Click to view available options
Quantity:
1 x 1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009907 1 x 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30009907 Supplier Toronto Research Chemicals Supplier No. TRCKIT89301X1ML
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 55-18-5
Molecular Formula C4 H10 N2 O
Quantity 1 x 1 mL
Synonym Ethanamine, N-ethyl-N-nitroso-, Diethylamine, N-nitroso- (6CI,8CI), N-Ethyl-N-nitrosoethanamine, DEN, DEN (mutagen), DENA, Diethylnitrosamide, Diethylnitrosamine, Diethylnitrosoamine, N,N-Diethylnitrosoamine, N-Nitroso-N,N-diethylamine, N-Nitrosodiethylamine, NDEA, NSC 132, Nitrosodiethylamine, N-Nitroso-diethylamine
InChI Formula InChI=1S/C4H10N2O/c1-3-6(4-2)5-7/h3-4H2,1-2H3
SMILES CCN(CC)N=O
IUPAC Name N,N-diethylnitrous amide
Molecular Weight (g/mol) 102.14
Formula Weight 102.0793
Concentration or Composition (by Analyte or Components) N-Nitroso-diethylamine 1 mg/mL
Chemical Name or Material N-Nitroso-diethylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.