missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodiethylamine, TRC

Product Code. 18090720
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18090720 1 g 1g
17939839 10 g 10g
18040820 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18090720 Supplier Toronto Research Chemicals Supplier No. TRCN5254651G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-18-5
Molecular Formula C4 H10 N2 O
Molecular Weight (g/mol) 102.14
Synonym Ethanamine, N-ethyl-N-nitroso-, Diethylamine, N-nitroso- (6CI,8CI), N-Ethyl-N-nitrosoethanamine, DEN, DEN (mutagen), DENA, Diethylnitrosamide, Diethylnitrosamine, Diethylnitrosoamine, N,N-Diethylnitrosoamine, N-Nitroso-N,N-diethylamine, N-Nitrosodiethylamine, NDEA, NSC 132, Nitrosodiethylamine, N-Nitroso-diethylamine
IUPAC Name N,N-diethylnitrous amide
SMILES CCN(CC)N=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H10N2O/c1-3-6(4-2)5-7/h3-4H2,1-2H3
Formula Weight 102.0793
Chemical Name or Material N-Nitroso-diethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.