missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodipropylamine, TRC

Product Code. 30004519
Change view
Click to view available options
Quantity:
250 mg
5 g
25 g
Unit Size:
250mg
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004519 25 g 25g
30011322 5 g 5g
18033710 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004519 Supplier Toronto Research Chemicals Supplier No. TRCN52548025G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-64-7
Molecular Formula C6 H14 N2 O
Molecular Weight (g/mol) 130.19
Synonym 1-Propanamine, N-nitroso-N-propyl-, Dipropylamine, N-nitroso- (6CI,7CI,8CI), N-Nitroso-N-propyl-1-propanamine, Dipropylnitrosamine, N,N-Dipropylnitrosamine, N-Nitroso-N-di-n-propylamine, N-Nitrosodi-n-propylamine, N-Nitrosodipropylamine, NSC 133, N-Nitroso-di-n-propylamine
IUPAC Name N,N-dipropylnitrous amide
SMILES CCCN(CCC)N=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H14N2O/c1-3-5-8(7-9)6-4-2/h3-6H2,1-2H3
Formula Weight 130.1106
Chemical Name or Material N-Nitroso-di-n-propylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.