missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosotris-(2-chloroethyl)urea (90%), TRC

Product Code. 30010315
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010315 25 mg 25mg
30003663 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010315 Supplier Toronto Research Chemicals Supplier No. TRCN54725025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69113-01-5
Molecular Formula C7 H12 Cl3 N3 O2
Molecular Weight (g/mol) 276.55
IUPAC Name 1,1,3-tris(2-chloroethyl)-3-nitrosourea
SMILES ClCCN(CCCl)C(=O)N(CCCl)N=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C7H12Cl3N3O2/c8-1-4-12(5-2-9)7(14)13(11-15)6-3-10/h1-6H2
Formula Weight 274.9995
Chemical Name or Material N-Nitrosotris-(2-chloroethyl)urea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.