Product Code. 10755642
Shop All Thermo Scientific Chemicals Products
Click to view available options
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
10755642
100 mL
AcroSeal™ Glass Bottle
100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
10755642
Supplier
Thermo Scientific Acros
Supplier No.
435631000
158.00 € 145.00 € / 100mL Save 13.00 € 8% Off
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
CAS: 111-84-2 | C9 H20 | 128.26 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS
111-84-2
CAS Min %
98.5
CAS Max %
100.0
Melting Point
-53.0°C
Color
Colorless
Density
0.7180g/mL
Boiling Point
151.0°C
Flash Point
31°C
Infrared Spectrum
Authentic
Assay Percent Range
98.5% min. (GC)
Packaging
AcroSeal™ Glass Bottle
Molecular Formula
C9 H20
Linear Formula
CH3 (CH2 )7 CH3
Refractive Index
1.4040 to 1.4060
Quantity
100 mL
Residue after Evaporation
0.0003% max.
Specific Gravity
0.718
Synonym
n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
Solubility Information
Solubility in water: insoluble. Other solubilities: soluble in alcohol, slightly soluble in benzene, acetone, chloroform
InChI Key
BKIMMITUMNQMOS-UHFFFAOYSA-N
SMILES
CCCCCCCCC
IUPAC Name
nonane
Molecular Weight (g/mol)
128.26
PubChem CID
8141
ChEBI
CHEBI:32892
Formula Weight
128.26
Percent Purity
99%
Physical Form
Liquid
Chemical Name or Material
n-Nonane
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?