missing translation for 'onlineSavingsMsg'
Learn More

n-Nonylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15445385
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445385 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15445385 Supplier Thermo Scientific Alfa Aesar Supplier No. H58604.AD
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 626207-32-7 | C9H19BrZn | 272.535 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 626207-32-7
Molecular Formula C9H19BrZn
Linear Formula CH3(CH2)8ZnBr
MDL Number MFCD09801447
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym n-nonylzinc bromide, nonylzinc bromide, n-nonylzinc bromide 0.5m in tetrahydrofuran, n-nonylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
InChI Key OXSAWWPTOCNGOR-UHFFFAOYSA-M
SMILES CCCCCCCC[CH2-].[Zn+]Br
IUPAC Name bromozinc(1+);nonane
Molecular Weight (g/mol) 272.535
PubChem CID 11311824
Formula Weight 272.53
Chemical Name or Material n-Nonylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.