missing translation for 'onlineSavingsMsg'
Learn More

N-Norbimatoprost, TRC

Product Code. 30138482
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138482 5 mg 5mg
30134516 10 mg 10mg
30006872 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138482 Supplier Toronto Research Chemicals Supplier No. TRCN6611105MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155205-89-3
Molecular Formula C23H33NO4
Molecular Weight (g/mol) 387.51
IUPAC Name (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(E,3S)-3-hydroxy-5-phenylpent-1-enyl]cyclopentyl]hept-5-enamide
SMILES NC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1\C=C\[C@@H](O)CCc2ccccc2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C23H33NO4/c24-23(28)11-7-2-1-6-10-19-20(22(27)16-21(19)26)15-14-18(25)13-12-17-8-4-3-5-9-17/h1,3-6,8-9,14-15,18-22,25-27H,2,7,10-13,16H2,(H2,24,28)/b6-1-,15-14+/t18-,19+,20+,21-,22+/m0/s1
Formula Weight 387.241
Chemical Name or Material N-Norbimatoprost
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.