missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)acetamide, 95%

Product Code. 10674822 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10674822 100 mL Glass bottle 100mL
10061810 25 mL Glass bottle 25mL
10564454 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10674822 Supplier Thermo Scientific Acros Supplier No. 156461000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10416-59-8 | C8H21NOSi2 | 203.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10416-59-8
Molecular Formula C8H21NOSi2
Molecular Weight (g/mol) 203.43
MDL Number MFCD00008270
InChI Key SIOVKLKJSOKLIF-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
PubChem CID 6913588
SMILES CC(O[Si](C)(C)C)=N[Si](C)(C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -8°C
Color Beige to Brown or Yellow
Density 0.8200g/mL
Boiling Point 71°C to 73°C (35.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3C[=NSi(CH3)3]OSi(CH3)3
Refractive Index 1.4165 to 1.4185
Quantity 100 mL
Fieser 01,61; 02,30; 03,23; 13,34
Specific Gravity 0.82
Solubility Information Solubility in water: decomposes
IUPAC Name trimethylsilyl N-(trimethylsilyl)ethanimidate
Formula Weight 203.43
Percent Purity 95%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material N, O-Bis(trimethylsilyl)acetamide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,O-Bis(trimethylsilyl)acetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.