missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS

Product Code. 11334988
Change view
Click to view available options
Quantity:
5 g
25 g
5 x 1 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334988 25 g 25g
11460640 5 x 1 g 1g
11324988 5 g 5g
11344988 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11334988 Supplier Thermo Scientific Alfa Aesar Supplier No. 043939.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.403 g/mol

N,O-Bis(trimethylsilyl)trifluoroacetamide is used as a derivatization reagent for gas chromatography and mass-spectrometry analysis of phenolic acids in fruits. It is also used as a reactant in the preparation of pyrimidinone ribosides and analogs which shows anti-tumor properties. It plays as an important role as a reagent for the regioselective desulfation of polysaccharide sulfates. Furthermore, it is used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.403
MDL Number MFCD00008269
InChI Key XCOBLONWWXQEBS-GHXNOFRVSA-N
Synonym bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
PubChem CID 9601896
IUPAC Name trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
SMILES C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C

Specifications

Melting Point -10°C
Color Colorless to Yellow
Density 0.97 g/mL
Boiling Point 145°C to 147°C
Flash Point 23°C (73°F)
Packaging Ampule
Refractive Index 1.384
Quantity 25 g
UN Number UN2924
Beilstein 3607620
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform.
Formula Weight 257.4
Physical Form Liquid
Chemical Name or Material N,O-Bis (trimethylsilyl)trifluoroacetamide, With 1% TMCS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.