missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS

Product Code. 11344988
Click to view available options
Quantity:
5 g
25 g
5 x 1 g
100 g
Unit Size:
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11344988

Brand: Thermo Scientific Alfa Aesar 043939.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.403 g/mol

N,O-Bis(trimethylsilyl)trifluoroacetamide is used as a derivatization reagent for gas chromatography and mass-spectrometry analysis of phenolic acids in fruits. It is also used as a reactant in the preparation of pyrimidinone ribosides and analogs which shows anti-tumor properties. It plays as an important role as a reagent for the regioselective desulfation of polysaccharide sulfates. Furthermore, it is used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.403
MDL Number MFCD00008269
InChI Key XCOBLONWWXQEBS-GHXNOFRVSA-N
Synonym bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
PubChem CID 9601896
IUPAC Name trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
SMILES C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C

Specifications

Melting Point -10°C
Color Colorless to Yellow
Density 0.97 g/mL
Boiling Point 145°C to 147°C
Flash Point 23°C (73°F)
Packaging Ampule
Refractive Index 1.384
Quantity 100 g
UN Number UN2924
Beilstein 3607620
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform.
Formula Weight 257.4
Physical Form Liquid
Chemical Name or Material N,O-Bis (trimethylsilyl)trifluoroacetamide, With 1% TMCS
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.