missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride

Product Code. 10330204 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10330204 100 g 100g
10005363 25 g 25g
11955861 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10330204 Supplier Thermo Scientific Supplier No. 389321000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25561-30-2 , 75-77-4
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.4
MDL Number MFCD00008269
InChI Key XCOBLONWWXQEBS-GHXNOFRVSA-N
Synonym bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
PubChem CID 9601896
IUPAC Name trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
SMILES C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C

Specifications

Color Colorless to Yellow
Density 0.9740g/mL
Boiling Point 145.0°C to 147.0°C
Flash Point 34°C
Packaging Glass bottle
Linear Formula CF3C[=NSi(CH3)3]OSi(CH3)3
Quantity 100 g
Specific Gravity 0.974
Solubility Information Solubility in water: hydrolysis
Formula Weight 257.4
Percent Purity ≥98.0% (excluding TMCS), 1.5% max. TMCS
Physical Form Liquid
Chemical Name or Material N, O-Bis(trimethylsilyl)trifluoroacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.