Learn More
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 10% TMCS
Description
N,O-Bis(trimethylsilyl)trifluoroacetamide is a powerful silylation reagent useful in GC and GC-MS for the derivatization of a wide range of functional groups under mild conditions. N,O. It is also used in HPLC- electrochemical detection assay of 8-oxo-deoxyguanosine and?8-oxo-guanine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Flash Point | 23°C (73°F) |
| Quantity | 5 g |
| UN Number | UN2924 |
| Beilstein | 3607620 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in water (hydrolyses),and chloroform. |
| Formula Weight | 257.4 |
| Physical Form | Liquid |
| Chemical Name or Material | N,O-Bis(trimethylsilyl)trifluoroacetamide, with 10% TMCS |
Sikkerhed og håndtering
- N,O-Bis(trimethylsilyl)trifluoroacetamide, with 10% TMCS
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : N,O-Bis(trimethylsilyl)trifluoroacetamide, Trimethylchlorosilane
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.