missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%

Product Code. 10790671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10790671 25 g Glass bottle 25g
10285930 5 g Glass bottle 5g
10409100 100 g Glass bottle 100g
10318493 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10790671 Supplier Thermo Scientific Acros Supplier No. 168000250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.39
MDL Number MFCD00008269
InChI Key XCOBLONWWXQEBS-GHXNOFRVSA-N
Synonym bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
PubChem CID 9601896
IUPAC Name trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
SMILES C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -10.0°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 45.0°C to 50.0°C (14.0 mmHg)
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CF3C[=NSi(CH3)3]OSi(CH3)3
Refractive Index 1.3829 to 1.3849
Quantity 25 g
Specific Gravity 0.96
Solubility Information Solubility in water: hydrolyses
Formula Weight 257.39
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material N, O-Bis(trimethylsilyl)trifluoroacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,O-Bis(trimethylsilyl)trifluoroacetamide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.