missing translation for 'onlineSavingsMsg'
Learn More

N,O-Dimethyl-Naltrexone Bromide, TRC

Product Code. 30383514
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30383514 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383514 Supplier Toronto Research Chemicals Supplier No. TRCD47499825MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,O-Dimethyl-Naltrexone Bromide
CAS 1005410-37-6
Molecular Formula C22H28BrNO4
Quantity 25 mg
Synonym (3R,4R,4aS,7aR,12bS)-3-(Cyclopropylmethyl)-4a-hydroxy-9-methoxy-3-methyl-7-oxo-2,3,4,4a,5,6,7,7a-octahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinolin-3-ium Bromide
InChI Formula InChI=1S/C22H28NO4.BrH/c1-23(12-13-3-4-13)10-9-21-18-14-5-6-16(26-2)19(18)27-20(21)15(24)7-8-22(21,25)17(23)11-14;/h5-6,13,17,20,25H,3-4,7-12H2,1-2H3;1H/q+1;/p-1/t17-,20+,21+,22-,23?;/m1./s1
SMILES [Br-].COc1ccc2C[C@@H]3[C@]4(O)CCC(=O)[C@@H]5Oc1c2[C@]45CC[N+]3(C)CC6CC6
IUPAC Name (4R,4aS,7aR,12bS)-3-(cyclopropylmethyl)-4a-hydroxy-9-methoxy-3-methyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-3-ium-7-one;bromide
Molecular Weight (g/mol) 450.37
Formula Weight 449.1202
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.