missing translation for 'onlineSavingsMsg'
Learn More

N,O-Dimethylhydroxylamine hydrochloride, 98%

Product Code. 11488067
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488067 100 g 100g
11468067 5 g 5g
11478067 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488067 Supplier Thermo Scientific Alfa Aesar Supplier No. A17469.22
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6638-79-5 | C2H8ClNO | 97.54 g/mol

N,O-Dimethylhydroxylamine hydrochloride is used as a reagent in the preparation of 2-acyloxazoles from 2-oxazolemagnesium chloride and N-methoxy-N-methyl-3-oxo-butyramide from diketene. It is also used in amide coupling reactions to form Weinreb amides, which are useful in the Weinreb ketone synthesis. In addition, it is used in the preparation of an inhibitor of the NLS-derived BC peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,O-Dimethylhydroxylamine hydrochloride is used as a reagent in the preparation of 2-acyloxazoles from 2-oxazolemagnesium chloride and N-methoxy-N-methyl-3-oxo-butyramide from diketene. It is also used in amide coupling reactions to form Weinreb amides, which are useful in the Weinreb ketone synthesis. In addition, it is used in the preparation of an inhibitor of the NLS-derived BC peptides.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6638-79-5
Molecular Formula C2H8ClNO
Molecular Weight (g/mol) 97.54
MDL Number MFCD00012485
InChI Key USZLCYNVCCDPLQ-UHFFFAOYSA-N
Synonym n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylamine hcl, n-methoxymethylamine hydrochloride, methanamine, n-methoxy-, hydrochloride, o,n-dimethylhydroxylamine hydrochloride, n-methoxymethanamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride
PubChem CID 81138
SMILES [H+].[Cl-].CNOC

Specifications

Melting Point 112°C to 115°C
Quantity 100 g
Beilstein 3650353
Sensitivity Hygroscopic
Solubility Information Soluble in water,dimethyl sulfoxide and methanol.
IUPAC Name N-methoxymethanamine;hydrochloride
Formula Weight 97.55
Percent Purity 98%
Chemical Name or Material N,O-Dimethylhydroxylamine hydrochloride
compliance-icons
Product Identifier
  • N,O-Dimethylhydroxylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N,O-Dimethylhydroxylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.