Product Code. 11388968
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
11388968
50 g
50g
11368968
10 g
10g
11378968
250 g
250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
11388968
Supplier
Thermo Scientific Alfa Aesar
Supplier No.
042460.18
Estimated Shipment: 05-10-2026Log in to see stock.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
CAS: 630-02-4 | C28H58 | 394.77 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
630-02-4
Molecular Formula
C28H58
Molecular Weight (g/mol)
394.77
MDL Number
MFCD00009355
InChI Key
ZYURHZPYMFLWSH-UHFFFAOYSA-N
Synonym
n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g
PubChem CID
12408
ChEBI
CHEBI:32943
IUPAC Name
octacosane
SMILES
CCCCCCCCCCCCCCCCCCCCCCCCCCCC
Specifications
Melting Point
61°C to 63°C
Boiling Point
440°C
Flash Point
227°C (441°F)
Quantity
50 g
Solubility Information
Soluble in chloroform,acetone,benzene,and toluene. Insoluble in water.
Formula Weight
394.77
Percent Purity
99%
Physical Form
Crystalline
Chemical Name or Material
n-Octacosane
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?