Learn More
n-Octadecane, 99%
Description
n-octadecane is used as solvent, lubricant, transformer oil and anti-corrosion agents. It is also used in paraffin as well as chemical intermediate in organic synthesis. It plays an important role in pheromones as a chemical messenger. N-Octadecane/ molecular sieve 5A composites find application as thermal storage material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 28°C |
| Density | 0.782 g/mL |
| Boiling Point | 317°C |
| Flash Point | 166°C (330°F) |
| Assay Percent Range | 99% |
| Odor | Odorless |
| Refractive Index | 1.439 |
| Quantity | 500 g |
| Solubility Information | Miscible with alcohol,acetone,ether and petroleum ether. Immiscible with water. |
| Formula Weight | 254.5 |
| Show More |
Safety and Handling
- n-Octadecane
- Danger
- Aspiration toxicity Category 1
- H304-May be fatal if swallowed and enters airways.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- MIXTURE LIST-Contains : Octadecane
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.