missing translation for 'onlineSavingsMsg'
Learn More

n-Octadecane, tech., 90%

Product Code. 11488390
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488390 5 kg 5kg
11498390 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11488390 Supplier Thermo Scientific Alfa Aesar Supplier No. 044301.A7
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 593-45-3 | C18H38 | 254.502 g/mol

n-octadecane is used as a solvent, a lubricant, transformer oil and an anti-corrosion agent. It is also used in paraffin and as a chemical intermediate in organic synthesis. It plays an important role in pheromones as a chemical messenger. N-Octadecane/ molecular sieve 5A composites find application as thermal storage material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-45-3
Molecular Formula C18H38
Molecular Weight (g/mol) 254.502
MDL Number MFCD00009007
InChI Key RZJRJXONCZWCBN-UHFFFAOYSA-N
Synonym n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group
PubChem CID 11635
ChEBI CHEBI:32926
IUPAC Name octadecane
SMILES CCCCCCCCCCCCCCCCCC

Specifications

Melting Point 28°C
Color White
Density 0.782 g/mL
Boiling Point 317°C
Flash Point 166°C (330°F)
Odor Odorless
Refractive Index 1.439
Quantity 5 kg
Solubility Information Miscible with alcohol,acetone,ether and petroleum ether. Immiscible with water.
Formula Weight 254.5
Percent Purity 90%
Grade Technical
Physical Form Crystalline Mass/Melt
Chemical Name or Material n-Octadecane
Show More Show Less
compliance-icons
Product Identifier
  • n-Octadecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: Octadecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.