missing translation for 'onlineSavingsMsg'
Learn More

n-Octadecyltrichlorosilane, 95%, cont. 5-10% branched isomers

Product Code. 11452368
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452368 50 g 50g
11462368 250 g 250g
11442368 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452368 Supplier Thermo Scientific Alfa Aesar Supplier No. A15732.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 112-04-9 | C18H37Cl3Si | 387.929 g/mol

n-Octadecyltrichlorosilane is used in semiconductor industry to form self-assembled monolayer thin films on silicon dioxide substrates. Used in molecular electronics, as thin insulating gates in Metal-Insulator-Semiconductor Field-Effect Transistors. It is also used in platinum catalyzed synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octadecyltrichlorosilane is used in semiconductor industry to form self-assembled monolayer thin films on silicon dioxide substrates. Used in molecular electronics, as thin insulating gates in Metal-Insulator-Semiconductor Field-Effect Transistors. It is also used in platinum catalyzed synthesis.

Solubility
Decomposes in water.

Notes
Moisture sensitive. Store in cool dry place in tightly closed container. Store under dry inert gas. Keep away from water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-04-9
Molecular Formula C18H37Cl3Si
Molecular Weight (g/mol) 387.929
MDL Number MFCD00000484
InChI Key PYJJCSYBSYXGQQ-UHFFFAOYSA-N
Synonym octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane
PubChem CID 8157
IUPAC Name trichloro(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 160°C to 162°C (3mmHg)
Melting Point ∼20°C
Quantity 50 g
Linear Formula CH3(CH2)17SiCl3
UN Number UN1800
Beilstein 1778847
Flash Point 189°C (372°F)
Solubility Information Decomposes in water.
Formula Weight 387.94
Percent Purity 95%
Odor Pungent
Refractive Index 1.46
Sensitivity Moisture sensitive
Density 0.984
Assay cont. 5 to 10% branched isomers
Chemical Name or Material n-Octadecyltrichlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Octadecyltrichlorosilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Octadecyltrichlorosilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.