missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Octanoyl-d15-glycine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-Octanoyl-d15-glycine |
| Molecular Formula | C10 D15 H4 N O3 |
| Quantity | 0.05 g |
| Synonym | N-(1-Oxooctyl-d15)glycine, N-Octanoylglycine-D15 (octanoyl-D15), Capryloylglycine-d15, N-Octanoyl-d15-glycine |
| InChI Formula | InChI=1 S/C10H19NO3/c1-2-3-4-5-6-7-9(12)11-8-10(13)14/h2-8H2,1H3,(H,11,12)(H,13,14)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)NCC(=O)O |
| IUPAC Name | 2-(2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecadeuteriooctanoylamino)acetic acid |
| Molecular Weight (g/mol) | 216.355 |
| Formula Weight | 216.231 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction