missing translation for 'onlineSavingsMsg'
Learn More

N-Octanoylglycine-13C2,15N, TRC

Product Code. 30416389
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416389 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416389 Supplier Toronto Research Chemicals Supplier No. TRCO23973710MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Octanoylglycine-13C2,15N
CAS 2483829-94-1
Molecular Formula C813C2H1915NO3
Quantity 10 mg
Synonym N-(1-Oxooctyl)glycine-3C2,15N, Capryloylglycine-3C2,15N, Lipacide C 8G-3C2,15N
InChI Formula InChI=1S/C10H19NO3/c1-2-3-4-5-6-7-9(12)11-8-10(13)14/h2-8H2,1H3,(H,11,12)(H,13,14)/i8+1,10+1,11+1
SMILES O=[13C](O)[13C]([H])([H])[15N]([H])C(CCCCCCC)=O
IUPAC Name octanoylglycine-13C2-15N
Molecular Weight (g/mol) 204.24
Formula Weight 204.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.