missing translation for 'onlineSavingsMsg'
Learn More

N-Octyl 4-Hydroxybenzoate, TRC

Product Code. 30410457
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410457 100 mg -
30425596 1 g 1g
30425368 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30410457 Supplier Toronto Research Chemicals Supplier No. TRCO148473100MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219-38-1
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.33
Synonym Benzoic acid, 4-hydroxy-, octyl ester (9CI, ACI), Benzoic acid, p-hydroxy-, octyl ester (6CI, 7CI, 8CI), n-Octyl 4-hydroxybenzoate, n-Octyl p-hydroxybenzoate, NSC 309820, Octyl 4-hydroxybenzoate, Octyl p-hydroxybenzoate, Octyl paraben, p-Hydroxybenzoic acid octyl ester
IUPAC Name octyl 4-hydroxybenzoate
SMILES CCCCCCCCOC(=O)c1ccc(O)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H22O3/c1-2-3-4-5-6-7-12-18-15(17)13-8-10-14(16)11-9-13/h8-11,16H,2-7,12H2,1H3
Formula Weight 250.1569
Chemical Name or Material N-Octyl 4-Hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.