missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl acetate, 98+%

Product Code. 11382506
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382506 50 g 50g
11392506 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382506 Supplier Thermo Scientific Alfa Aesar Supplier No. L06405.18
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 112-14-1 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-14-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00009562
InChI Key YLYBTZIQSIBWLI-UHFFFAOYSA-N
Synonym n-octyl acetate, acetic acid, octyl ester, caprylyl acetate, 1-octyl acetate, acetic acid octyl ester, octyl ethanoate, n-octanyl acetate, n-octyl ethanoate, 1-octanol acetate, acetate c-8
PubChem CID 8164
ChEBI CHEBI:87495
IUPAC Name octyl acetate
SMILES CCCCCCCCOC(C)=O

Specifications

Density 0.868
Boiling Point 210°C to 211°C
Flash Point 86°C (186°F)
Linear Formula CH3CO2(CH2)7CH3
Refractive Index 1.419
Quantity 50 g
Beilstein 1754554
Formula Weight 172.27
Percent Purity ≥98%
Chemical Name or Material n-Octyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.