missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl-beta-D-thioglucopyranoside, 98+%

Product Code. 15483689
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483689 10 g 10g
15493689 2.5 g 2.5g
15473689 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15483689 Supplier Thermo Scientific Alfa Aesar Supplier No. J61028.09
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

Non-ionic detergent used for the solubilization of membrane proteins | CAS: 85618-21-9 | C14H28O5S | 308.433 g/mol

n-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.

Solubility
Soluble in ethanol, water or methanol

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85618-21-9
Molecular Formula C14H28O5S
Molecular Weight (g/mol) 308.433
MDL Number MFCD00012189
InChI Key CGVLVOOFCGWBCS-DGTMBMJNSA-N
Synonym octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside
PubChem CID 53720324
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol
SMILES CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O

Specifications

Melting Point 128°C
Color White
Quantity 10 g
Beilstein 4182783
Solubility Information Soluble in ethanol, water or methanol
Formula Weight 308.44
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material n-Octyl-beta-D-thioglucopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.