missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl butyrate, 97%

Product Code. 11491547
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491547 50 g 50g
11401557 250 g 250g
11481547 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491547 Supplier Thermo Scientific Alfa Aesar Supplier No. B24978.18
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-39-4 | C12H24O2 | 200.322 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-39-4
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00048939
InChI Key PWLNAUNEAKQYLH-UHFFFAOYSA-N
Synonym octyl butyrate, butanoic acid, octyl ester, n-octyl butyrate, butyric acid, octyl ester, n-octyl n-butyrate, n-octyl butanoate, unii-5yeu4o369l, butyric acid, octyl ester 8ci, octyl butyrate natural, fema no. 2807
PubChem CID 61030
IUPAC Name octyl butanoate
SMILES CCCCCCCCOC(=O)CCC

Specifications

Quantity 50 g
Formula Weight 172.27
Percent Purity 97%
Chemical Name or Material n-Octyl butyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.